Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Hexenyl Acetate 97.0+%, TCI America™

Click to view available options
Quantity:
25mL
250mL
2497-18-9
C8H14O2
142.198
MFCD00009474
HRHOWZHRCRZVCU-AATRIKPKSA-N
z-hex-2-enyl acetate,unii-pt792bq57d,2-hexen-1-ol, acetate, z,z-2-hexenyl acetate,fema no. 2564,2-hexen-1-yl acetate, trans,2-hexen-1-ol, acetate, 2e,2-hexenyl acetate, trans,hexenyl acetate, 2z
2733294
[(E)-hex-2-enyl] acetate
CCCC=CCOC(=O)C
2497-18-9
C8H14O2
142.198
MFCD00009474
HRHOWZHRCRZVCU-AATRIKPKSA-N
z-hex-2-enyl acetate,unii-pt792bq57d,2-hexen-1-ol, acetate, z,z-2-hexenyl acetate,fema no. 2564,2-hexen-1-yl acetate, trans,2-hexen-1-ol, acetate, 2e,2-hexenyl acetate, trans,hexenyl acetate, 2z
2733294
[(E)-hex-2-enyl] acetate
CCCC=CCOC(=O)C
Boiling Point 165°C
Color Colorless
Quantity 250mL
Formula Weight 142.20
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material trans-2-Hexenyl Acetate

EINECSNumber : (2)-0760&(2)-2533

RTECSNumber : MP8425000

TSCA : Yes