Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ trans-2-Octenal, 95%

Click to view available options
Quantity:
5g
25g
2548-87-0
C8H14O
126.199
MFCD00007011
LVBXEMGDVWVTGY-VOTSOKGWSA-N
trans-2-octenal,e-oct-2-enal,e-2-octenal,2-octenal,trans-2-octen-1-al,e-2-octen-1-al,2-octen-1-al,2-octenel,2-octenal, e,2-octenal, 2e
5283324
CHEBI:61748
(E)-oct-2-enal
CCCCCC=CC=O
2548-87-0
C8H14O
126.199
MFCD00007011
LVBXEMGDVWVTGY-VOTSOKGWSA-N
trans-2-octenal,e-oct-2-enal,e-2-octenal,2-octenal,trans-2-octen-1-al,e-2-octen-1-al,2-octen-1-al,2-octenel,2-octenal, e,2-octenal, 2e
5283324
CHEBI:61748
(E)-oct-2-enal
CCCCCC=CC=O
Applications
Used in flavoring tangerine, mandarin, cherry, melon and orange and also as a fragrances agent. It causes defects for locomotion in green fluorescent protein (GFP). It is an antimicrobial agent that destroys fungi by suppressing their ability to grow or reproduce.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and fixed oils.

Notes
Air Sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Refractive Index 1.45
Odor Pleasant
Sensitivity Air Sensitive
Density 0.84
Flash Point 71°C (160°F)
Quantity 5g
Boiling Point 84°C to 86°C (19mmHg)
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol and fixed oils.
Formula Weight 126.2
Percent Purity 95%
Chemical Name or Material trans-2-Octenal
Show More Show Less

GHS H Statement
H315-H317-H227
Causes skin irritation.
May cause an allergic skin reaction.
Combustible liquid.

P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P370+P378q-P501c

H227-H315-H317-H319

EINECSNumber : 219-833-8

TSCA : Yes

Recommended Storage : Ambient temperatures