Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Octene 98%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
Packaging:
Glass bottle
13389-42-9
C8H16
112.216
MFCD00009532
ILPBINAXDRFYPL-HWKANZROSA-N
trans-2-octene,2-octene,e-2-octene,e-oct-2-ene,2-octene, e,2-octene, 2e,unii-g13g1yr8yw,oct-2-ene,g13g1yr8yw,octene-2
5364448
(E)-oct-2-ene
CCCCCC=CC
13389-42-9
C8H16
112.216
MFCD00009532
ILPBINAXDRFYPL-HWKANZROSA-N
trans-2-octene,2-octene,e-2-octene,e-oct-2-ene,2-octene, e,2-octene, 2e,unii-g13g1yr8yw,oct-2-ene,g13g1yr8yw,octene-2
5364448
(E)-oct-2-ene
CCCCCC=CC
Infrared Spectrum Authentic
Density 0.7180g/mL
Packaging Glass bottle
Quantity 5g
Color Colorless
Refractive Index 1.4120 to 1.4140 (20°C, 589nm)
NMR Complies
Assay Percent Range 98%
Linear Formula CH3(CH2)4CH=CHCH3
Formula Weight 112.21
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material trans-2-Octene, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 236-463-2