Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Pentene 95%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
646-04-8
C5H10
70.135
MFCD00009384
QMMOXUPEWRXHJS-HWKANZROSA-N
trans-2-pentene,e-2-pentene,2-pentene,3-pentene,2-pentene, e,sym-methylethylethylene,e-pent-2-ene,beta-n-amylene,trans-beta-amylene,2-pentene, 2e
5326161
(E)-pent-2-ene
CCC=CC
646-04-8
C5H10
70.135
MFCD00009384
QMMOXUPEWRXHJS-HWKANZROSA-N
trans-2-pentene,e-2-pentene,2-pentene,3-pentene,2-pentene, e,sym-methylethylethylene,e-pent-2-ene,beta-n-amylene,trans-beta-amylene,2-pentene, 2e
5326161
(E)-pent-2-ene
CCC=CC
Color Colorless to Yellow
Quantity 5g
Refractive Index 1.3785 to 1.3805 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Density 0.6400g/mL
Assay Percent Range 95%
Linear Formula C2H5CH=CHCH3
Beilstein 01, 210
Merck Index 15, 7235
Formula Weight 70.13
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material trans-2-Pentene, 95%
Show More Show Less

GHS H Statement:
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-461-4

RTECSNumber : SB2184500

TSCA : TSCA