Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Phenylcyclopropane-1-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
939-90-2
C10H10O2
162.188
MFCD00001292
AHDDRJBFJBDEPW-DTWKUNHWSA-N
trans-2-phenylcyclopropanecarboxylic acid,trans-2-phenyl-1-cyclopropanecarboxylic acid,1r,2r-2-phenylcyclopropanecarboxylic acid,trans-2-phenylcyclopropane-1-carboxylic acid,2-phcpcooh,1r,2r-2-phenylcyclopropane-1-carboxylic acid,cyclopropanecarboxylic acid, 2-phenyl-, 1r,2r-rel,rac trans-2-phenylcyclopropanecarboxylic-d5 acid,pubchem22341,trans-2-phenylcyclopropancarboxylic acid
237413
CHEBI:79920
(1R,2R)-2-phenylcyclopropane-1-carboxylic acid
C1C(C1C(=O)O)C2=CC=CC=C2
939-90-2
C10H10O2
162.188
MFCD00001292
AHDDRJBFJBDEPW-DTWKUNHWSA-N
trans-2-phenylcyclopropanecarboxylic acid,trans-2-phenyl-1-cyclopropanecarboxylic acid,1r,2r-2-phenylcyclopropanecarboxylic acid,trans-2-phenylcyclopropane-1-carboxylic acid,2-phcpcooh,1r,2r-2-phenylcyclopropane-1-carboxylic acid,cyclopropanecarboxylic acid, 2-phenyl-, 1r,2r-rel,rac trans-2-phenylcyclopropanecarboxylic-d5 acid,pubchem22341,trans-2-phenylcyclopropancarboxylic acid
237413
CHEBI:79920
(1R,2R)-2-phenylcyclopropane-1-carboxylic acid
C1C(C1C(=O)O)C2=CC=CC=C2
Infrared Spectrum Authentic
Melting Point 86.0°C to 92.0°C
Quantity 5g
Color Beige or Yellow
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula C6H5C3H4CO2H
Formula Weight 162.19
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material trans-2-Phenylcyclopropane-1-carboxylic acid
Show More Show Less