Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Phenylcyclopropylamine hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
1986-47-6
C9H11N·HCl
169.66
MFCD00063602
ZPEFMSTTZXJOTM-VTLYIQCISA-N
tranylcypromine hydrochloride,trans-2-phenylcyclopropanamine hydrochloride,trans-2-phenylcyclopropylamine hydrochloride,1r,2s-2-phenyl-cyclopropylamine hydrochloride,1r,2s-2-phenylcyclopropan-1-amine hydrochloride,tranylcypromine hcl,1r,2s-2-phenylcyclopropanamine hydrochloride,trans-2-phenyl-cyclopropylamine hydrochloride,dsstox_cid_27761,dsstox_rid_82538
12345947
(1R,2R)-2-phenylcyclopropan-1-amine;hydrochloride
C1C(C1N)C2=CC=CC=C2.Cl
1986-47-6
C9H11N·HCl
169.66
MFCD00063602
ZPEFMSTTZXJOTM-VTLYIQCISA-N
tranylcypromine hydrochloride,trans-2-phenylcyclopropanamine hydrochloride,trans-2-phenylcyclopropylamine hydrochloride,1r,2s-2-phenyl-cyclopropylamine hydrochloride,1r,2s-2-phenylcyclopropan-1-amine hydrochloride,tranylcypromine hcl,1r,2s-2-phenylcyclopropanamine hydrochloride,trans-2-phenyl-cyclopropylamine hydrochloride,dsstox_cid_27761,dsstox_rid_82538
12345947
(1R,2R)-2-phenylcyclopropan-1-amine;hydrochloride
C1C(C1N)C2=CC=CC=C2.Cl
Melting Point 162°C to 169°C
Quantity 1g
Infrared Spectrum Authentic
Color White to Beige
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula C6H5C3H4NH2·HCl
Beilstein 12, III, 2797
Merck Index 14, 9573
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ethyl acetate,diethyl ether
Formula Weight 169.66
Physical Form Powder or Chunks
Percent Purity ≥96.0%
Chemical Name or Material trans-2-Phenylcyclopropylamine hydrochloride
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger