Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
trans-3-Chloroacrylic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
trans-3-Chloroacrylic acid is a chlorinated carboxylic acid for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-3-Chloroacrylic acid is a chlorinated carboxylic acid for proteomics research.
Solubility
Soluble in water.
Notes
Store at 4°C. Store away from oxidizing agent, bases.
trans-3-Chloroacrylic acid is a chlorinated carboxylic acid for proteomics research.
Solubility
Soluble in water.
Notes
Store at 4°C. Store away from oxidizing agent, bases.
Specifications
Melting Point | 81°C to 85°C |
Quantity | 250 mg |
UN Number | UN3261 |
Beilstein | 1720279 |
Solubility Information | Soluble in water. |
SMILES | C(=CCl)C(=O)O |
Molecular Weight (g/mol) | 106.505 |
Formula Weight | 106.51 |
Percent Purity | 99% |
Chemical Name or Material | trans-3-Chloroacrylic acid |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 219-070-0 |
RTECSNumber | AS5960000 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction