Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-3-chloroacrylic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2345-61-1
C3H3ClO2
106.51
MFCD00064237
MHMUCYJKZUZMNJ-OWOJBTEDSA-N
trans-3-chloroacrylic acid,e-3-chloroacrylic acid,3-chloroacrylic acid,3-chloroprop-2-enoic acid,acrylic acid, 3-chloro-, trans,acrylic acid, 3-chloro-, e,trans-3-chloropropenoic acid,trans-beta-chloroacrylic acid,2-propenoic acid, 3-chloro-, e,ccris 3546
638124
(E)-3-chloroprop-2-enoic acid
C(=CCl)C(=O)O
2345-61-1
C3H3ClO2
106.51
MFCD00064237
MHMUCYJKZUZMNJ-OWOJBTEDSA-N
trans-3-chloroacrylic acid,e-3-chloroacrylic acid,3-chloroacrylic acid,3-chloroprop-2-enoic acid,acrylic acid, 3-chloro-, trans,acrylic acid, 3-chloro-, e,trans-3-chloropropenoic acid,trans-beta-chloroacrylic acid,2-propenoic acid, 3-chloro-, e,ccris 3546
638124
(E)-3-chloroprop-2-enoic acid
C(=CCl)C(=O)O
Color Beige-Gray to Gray
Infrared Spectrum Authentic
Melting Point 82°C to 85°C
Quantity 1g
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula ClCH=CHCO2H
Beilstein 02, 400
Formula Weight 106.51
Physical Form Powder or Flakes
Percent Purity 99%
Chemical Name or Material trans-3-chloroacrylic acid
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 219-070-