Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-3-Hexene, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13269-52-8
C6H12
84.16
ZQDPJFUHLCOCRG-AATRIKPKSA-N
trans-3-hexene,3-hexene,e-3-hexene,3-hexene, e,3-hexene, 3e,unii-3co1lux1ar,e-hex-3-ene,3co1lux1ar,alkenes, c6,cis-3-hexene
638066
(E)-hex-3-ene
CCC=CCC
13269-52-8
C6H12
84.16
ZQDPJFUHLCOCRG-AATRIKPKSA-N
trans-3-hexene,3-hexene,e-3-hexene,3-hexene, e,3-hexene, 3e,unii-3co1lux1ar,e-hex-3-ene,3co1lux1ar,alkenes, c6,cis-3-hexene
638066
(E)-hex-3-ene
CCC=CCC
Flash Point −12°C
Specific Gravity 0.677
Packaging Glass bottle
Melting Point -113.0°C
Boiling Point 67.0°C
Refractive Index 1.3935 to 1.3955
Density 0.6770g/mL
Quantity 5g
Assay Percent Range 98%
Linear Formula C2H5CH=CHC2H5
Beilstein 01,II,192
Formula Weight 84.16
Percent Purity 98%
Chemical Name or Material trans-3-Hexene
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger