Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-3-Hexene, 98%, Acros Organics™

Click to view available options
Quantity:
5mL
13269-52-8
C6H12
84.162
ZQDPJFUHLCOCRG-AATRIKPKSA-N
trans-3-hexene,3-hexene,e-3-hexene,3-hexene, e,3-hexene, 3e,unii-3co1lux1ar,e-hex-3-ene,3co1lux1ar,alkenes, c6,cis-3-hexene
638066
(E)-hex-3-ene
CCC=CCC
13269-52-8
C6H12
84.162
ZQDPJFUHLCOCRG-AATRIKPKSA-N
trans-3-hexene,3-hexene,e-3-hexene,3-hexene, e,3-hexene, 3e,unii-3co1lux1ar,e-hex-3-ene,3co1lux1ar,alkenes, c6,cis-3-hexene
638066
(E)-hex-3-ene
CCC=CCC
Boiling Point 67.0°C
Quantity 5mL
Density 0.6770g/mL
Specific Gravity 0.677
Melting Point -113.0°C
Refractive Index 1.3935 to 1.3955
Flash Point −12°C
Linear Formula C2H5CH=CHC2H5
Formula Weight 84.16
Chemical Name or Material trans-3-Hexene