Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-3-Hexenoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
1577-18-0
C6H10O2
114.144
MFCD00002786
XXHDAWYDNSXJQM-ONEGZZNKSA-N
trans-3-hexenoic acid,hydrosorbic acid,3-hexenoic acid,e-hex-3-enoic acid,3-hexenoic acid, 3e,e-3-hexenoic acid,3-hexenoic acid, e,trans-hex-3-enoic acid,unii-9b3n069edg,fema no. 3170
5282708
CHEBI:49285
(E)-hex-3-enoic acid
CCC=CCC(=O)O
1577-18-0
C6H10O2
114.144
MFCD00002786
XXHDAWYDNSXJQM-ONEGZZNKSA-N
trans-3-hexenoic acid,hydrosorbic acid,3-hexenoic acid,e-hex-3-enoic acid,3-hexenoic acid, 3e,e-3-hexenoic acid,3-hexenoic acid, e,trans-hex-3-enoic acid,unii-9b3n069edg,fema no. 3170
5282708
CHEBI:49285
(E)-hex-3-enoic acid
CCC=CCC(=O)O
Density 0.9650g/mL
Color Colorless to Yellow
Quantity 50g
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4370 to 1.4450 (20°C, 589nm)
Assay Percent Range 97%
Linear Formula C2H5CH=CHCH2CO2H
Beilstein 02, 435
Formula Weight 114.14
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material trans-3-Hexenoic acid, 97%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 216-417-8

TSCA : TSCA