Alfa Aesar™ trans-3-Octene, 97%

Manufacturer: Alfa Aesar™ 4315404

Quantity 2g

 View more versions of this product

Catalog No. AA4315404

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14919-01-8
    Molecular Formula C8H16
    Formula Weight 112.22
    MDL Number MFCD00039971
    UN Number UN3295
    Chemical Name or Material trans-3-Octene

    Specifications

    Formula Weight 112.22
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Refractive Index 1.4130
    CAS 14919-01-8
    UN Number UN3295
    Boiling Point 121° to 222°C
    Health Hazard 1 Danger
    Flash Point 13°C (57°F)
    Odor Characteristic
    Physical Form Liquid
    Molecular Formula C8H16
    Density 0.716g/mL
    Quantity 2g
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapor.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    MDL Number MFCD00039971
    Chemical Name or Material trans-3-Octene
    Assay 97%

    GHS H Statement
    H225
    Highly flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit