Alfa Aesar™ trans-4-(4-n-Propylcyclohexyl)cyclohexanone, 97%

Manufacturer: Alfa Aesar™ H5539103

Quantity 1g

 View more versions of this product

Catalog No. AAH5539103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 82832-73-3
    Molecular Formula C15H26O
    Formula Weight 222.37
    MDL Number MFCD04038797
    Chemical Name or Material trans-4-(4-n-Propylcyclohexyl)cyclohexanone

    Specifications

    Formula Weight 222.37
    Molecular Formula C15H26O
    MDL Number MFCD04038797
    CAS 82832-73-3
    Quantity 1g
    Chemical Name or Material trans-4-(4-n-Propylcyclohexyl)cyclohexanone
    Assay 97%

    Description

    Applications
    Used as pharmaceutical intermediate.

    Notes
    Keep container tightly closed. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit