Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4,5'-Dinitrodibenzo-18-crown-6, 98%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
32082-45-4
C20H22N2O10
450.4
JLRXOTNWTNNRCW-UHFFFAOYSA-N
dibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecin, 6,7,9,10,17,18,20,21-octahydro-2,13-dinitro,2,13-dinitro-6,7,9,10,17,18,20,21-octahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine,3,3'-dinitrobenzo-18-crown-6,trans-di nitrobenzo-18-crown-6,dinitrodibenzo-18-crown-6,20,24-dinitro-dibenzo-18-crown-6,3,4-4-nitro-2,1-phenylenebis oxyethyleneoxyethyleneoxy-1-nitrobenzene,4',5-dinitrodibenzo b,k-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene,11,24-dinitro-2,5,8,15,18,21-hexaoxatricyclo 20.4.0.0?, 1 ? hexacosa-1 22 ,9,11,13,23,25-hexaene,11,24-dinitro-2,5,8,15,18,21-hexaoxatricyclo 20.4.0.0^ 9,14 hexacosa-1 22 ,9 14 ,10,12,23,25-hexaene
360968
C1COC2=C(C=C(C=C2)[N+](=O)[O-])OCCOCCOC3=C(C=C(C=C3)[N+](=O)[O-])OCCO1
32082-45-4
C20H22N2O10
450.4
JLRXOTNWTNNRCW-UHFFFAOYSA-N
dibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecin, 6,7,9,10,17,18,20,21-octahydro-2,13-dinitro,2,13-dinitro-6,7,9,10,17,18,20,21-octahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine,3,3'-dinitrobenzo-18-crown-6,trans-di nitrobenzo-18-crown-6,dinitrodibenzo-18-crown-6,20,24-dinitro-dibenzo-18-crown-6,3,4-4-nitro-2,1-phenylenebis oxyethyleneoxyethyleneoxy-1-nitrobenzene,4',5-dinitrodibenzo b,k-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene,11,24-dinitro-2,5,8,15,18,21-hexaoxatricyclo 20.4.0.0?, 1 ? hexacosa-1 22 ,9,11,13,23,25-hexaene,11,24-dinitro-2,5,8,15,18,21-hexaoxatricyclo 20.4.0.0^ 9,14 hexacosa-1 22 ,9 14 ,10,12,23,25-hexaene
360968
C1COC2=C(C=C(C=C2)[N+](=O)[O-])OCCOCCOC3=C(C=C(C=C3)[N+](=O)[O-])OCCO1
Chelation/Complexation reagent
Quantity 1g
Packaging Glass bottle
Assay Percent Range 98%
Formula Weight 450.4
Chemical Name or Material trans-4, 5'-Dinitrodibenzo-18-crown-6