Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Acetamidocyclohexanol 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
27489-60-7
C8H15NO2
157.213
MFCD00051422
HWAFCRWGGRVEQL-UHFFFAOYSA-N
90074
N-(4-hydroxycyclohexyl)acetamide
CC(=O)NC1CCC(CC1)O
27489-60-7
C8H15NO2
157.213
MFCD00051422
HWAFCRWGGRVEQL-UHFFFAOYSA-N
90074
N-(4-hydroxycyclohexyl)acetamide
CC(=O)NC1CCC(CC1)O
Quantity 25g
Color Undesignated
Melting Point 170°C
Formula Weight 157.21
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material trans-4-Acetamidocyclohexanol