Alfa Aesar™ trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride, 97%

Manufacturer: Alfa Aesar™ H52566MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52566MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 334999-32-5
    Molecular Formula C11H20N2O4·HCl
    Formula Weight 280.75
    MDL Number MFCD03787929
    Chemical Name or Material trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride

    Specifications

    Formula Weight 280.75
    Molecular Formula C11H20N2O4·HCl
    MDL Number MFCD03787929
    CAS 334999-32-5
    Quantity 250mg
    Chemical Name or Material trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride
    Assay 97%

    Description

    Applications
    trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride acts as a pharmaceutical intermediate. It is used to prepare N-Boc-protected proline methyl ester of calix[4]arene, which is efficiently catalyzes the enantioselective aldol reaction.

    Notes
    Store in a cool place. Moisture sensitive. Incompatible with oxidizing agents and acids.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit