Alfa Aesar™ trans-4-Aminocyclohexanol, 98+%

Manufacturer: Alfa Aesar™ B2236522

Quantity 100g

 View more versions of this product

Catalog No. AAB2236522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 27489-62-9
    Molecular Formula C6H13NO
    Formula Weight 115.18
    MDL Number MFCD00067698
    UN Number UN3259
    Synonym trans-4-Hydroxycyclohexylamine
    Chemical Name or Material trans-4-Aminocyclohexanol

    Specifications

    Formula Weight 115.18
    Melting Point 110° to 113°C
    Quantity 100g
    Molecular Formula C6H13NO
    MDL Number MFCD00067698
    CAS 27489-62-9
    UN Number UN3259
    Synonym trans-4-Hydroxycyclohexylamine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
    Chemical Name or Material trans-4-Aminocyclohexanol
    Assay ≥98%

    Description

    Applications
    trans-4-Aminocyclohexanol is used as raw material in organic synthesis and is an important intermediate in the synthesis of drugs such as Ambroxol hydrochloride. It can react with butyric acid-(2-chloro-ethyl ester) to get butyric acid 4-amino-cyclohexyl ester. This reaction needs catalytic agent Aspergillus niger lipase and solvent 2-methyl-butan-2-ol.

    Solubility
    Soluble in water.

    Notes
    Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit