Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ trans-4-Aminocyclohexanol, 98+%

Click to view available options
Quantity:
5g
25g
100g
27489-62-9
C6H13NO
115.176
MFCD00067698
IMLXLGZJLAOKJN-UHFFFAOYSA-N
trans-4-aminocyclohexanol,4-aminocyclohexanol,cis-4-aminocyclohexanol,cis-4-amino-cyclohexanol,cyclohexanol, 4-amino-, trans,1r,4r-4-aminocyclohexan-1-ol,cyclohexanol, 4-amino,trans-4-hydroxycyclohexylamine,1r,4r-4-aminocyclohexanol,aminocyclohexanol trans-4
81293
4-aminocyclohexan-1-ol
C1CC(CCC1N)O
27489-62-9
C6H13NO
115.176
MFCD00067698
IMLXLGZJLAOKJN-UHFFFAOYSA-N
trans-4-aminocyclohexanol,4-aminocyclohexanol,cis-4-aminocyclohexanol,cis-4-amino-cyclohexanol,cyclohexanol, 4-amino-, trans,1r,4r-4-aminocyclohexan-1-ol,cyclohexanol, 4-amino,trans-4-hydroxycyclohexylamine,1r,4r-4-aminocyclohexanol,aminocyclohexanol trans-4
81293
4-aminocyclohexan-1-ol
C1CC(CCC1N)O
Applications
trans-4-Aminocyclohexanol is used as raw material in organic synthesis and is an important intermediate in the synthesis of drugs such as Ambroxol hydrochloride. It can react with butyric acid-(2-chloro-ethyl ester) to get butyric acid 4-amino-cyclohexyl ester. This reaction needs catalytic agent Aspergillus niger lipase and solvent 2-methyl-butan-2-ol.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Melting Point 110°C to 113°C
Quantity 25g
UN Number UN3259
Formula Weight 115.18
Percent Purity ≥98%
Chemical Name or Material trans-4-Aminocyclohexanol

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.