Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ trans-4-Aminocyclohexanol hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
100g
50910-54-8
C6H14ClNO
151.634
MFCD00012566
RKTQEVMZBCBOSB-UHFFFAOYSA-N
trans-4-aminocyclohexanol hydrochloride,cis-4-aminocyclohexanol hydrochloride,4-aminocyclohexanol hydrochloride,cis-4-aminocyclohexanol hcl,4-aminocyclohexan-1-ol hydrochloride,cis-4-amino-cyclohexanol hydrochloride,4-amino-cyclohexanol hydrochloride,1r,4r-4-aminocyclohexan-1-ol hydrochloride,cyclohexanol, 4-amino-, hydrochloride,trans-4-aminocyclohexanol hcl
522619
4-aminocyclohexan-1-ol;hydrochloride
C1CC(CCC1N)O.Cl
50910-54-8
C6H14ClNO
151.634
MFCD00012566
RKTQEVMZBCBOSB-UHFFFAOYSA-N
trans-4-aminocyclohexanol hydrochloride,cis-4-aminocyclohexanol hydrochloride,4-aminocyclohexanol hydrochloride,cis-4-aminocyclohexanol hcl,4-aminocyclohexan-1-ol hydrochloride,cis-4-amino-cyclohexanol hydrochloride,4-amino-cyclohexanol hydrochloride,1r,4r-4-aminocyclohexan-1-ol hydrochloride,cyclohexanol, 4-amino-, hydrochloride,trans-4-aminocyclohexanol hcl
522619
4-aminocyclohexan-1-ol;hydrochloride
C1CC(CCC1N)O.Cl
Melting Point 225°C to 230°C
Quantity 5g
Beilstein 3909294
Formula Weight 151.64
Percent Purity 97%
Chemical Name or Material trans-4-Aminocyclohexanol hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning