Learn More
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals
Description
It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.
Solubility
Soluble in water (167 mg/ml), ethanol (<1mg/mL at 25°C). Very slightly soluble in ether, DMSO (<1mg/mL at 25°C), and alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | >300°C |
Beilstein | 2207452 |
Merck Index | 14,9569 |
Solubility Information | Soluble in water (167mg/ml),ethanol (<1mg/ml at 25°C). Very slightly soluble in ether,DMSO (<1mg/ml at 25°C),and alcohol. |
Formula Weight | 157.21 |
Percent Purity | 97% |
Quantity | 10 g |
Chemical Name or Material | trans-4-(Aminomethyl)cyclohexanecarboxylic acid |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 214-818-2 |
RTECSNumber | GU8400000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.