Alfa Aesar™ trans-4-Benzyl-L-proline hydrochloride, 95%

Manufacturer: Alfa Aesar™ H5204103

Quantity 1g

 View more versions of this product

Catalog No. AAH5204103

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C12H15NO2·HCl
    Formula Weight 241.72
    MDL Number MFCD15143631
    Chemical Name or Material trans-4-Benzyl-L-proline hydrochloride

    Specifications

    Formula Weight 241.72
    Molecular Formula C12H15NO2·HCl
    MDL Number MFCD15143631
    Quantity 1g
    Chemical Name or Material trans-4-Benzyl-L-proline hydrochloride
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit