Alfa Aesar™ trans-4-Benzyl-N-Boc-L-proline, 95%

Manufacturer: Alfa Aesar™ H52135MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52135MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 153074-95-4
    Molecular Formula C17H23NO4
    Formula Weight 305.37
    MDL Number MFCD07784045
    Chemical Name or Material trans-4-Benzyl-N-Boc-L-proline

    Specifications

    Formula Weight 305.37
    Molecular Formula C17H23NO4
    MDL Number MFCD07784045
    CAS 153074-95-4
    Melting Point 100° to 102°C
    Quantity 250mg
    Chemical Name or Material trans-4-Benzyl-N-Boc-L-proline
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit