Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
34771-04-5
C15H19NO4
277.32
MFCD01550737
ZVMICQYOGWAOSU-UHFFFAOYSA-N
trans-4-carbobenzoxyamino cyclohexanecarboxylic acid,trans-4-cbz-amino cyclohexanecarboxylic acid,4-benzyloxycarbonylamino-cyclohexanecarboxylic acid,trans-4-benzyloxycarbonylaminocyclohexanecarboxylic acid,cis-4-benzyloxycarbonylaminocyclohexanecarboxylic acid,cis-4-benzyloxycarbonylaminocyclohexane-carboxylic acid,4-benzyloxy carbonyl amino cyclohexanecarboxylic acid,4-phenylmethoxycarbonylamino cyclohexane-1-carboxylic acid,trans-4-benzyloxy carbonyl amino cyclohexanecarboxylic acid
285247
4-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
C1CC(CCC1C(=O)O)NC(=O)OCC2=CC=CC=C2
34771-04-5
C15H19NO4
277.32
MFCD01550737
ZVMICQYOGWAOSU-UHFFFAOYSA-N
trans-4-carbobenzoxyamino cyclohexanecarboxylic acid,trans-4-cbz-amino cyclohexanecarboxylic acid,4-benzyloxycarbonylamino-cyclohexanecarboxylic acid,trans-4-benzyloxycarbonylaminocyclohexanecarboxylic acid,cis-4-benzyloxycarbonylaminocyclohexanecarboxylic acid,cis-4-benzyloxycarbonylaminocyclohexane-carboxylic acid,4-benzyloxy carbonyl amino cyclohexanecarboxylic acid,4-phenylmethoxycarbonylamino cyclohexane-1-carboxylic acid,trans-4-benzyloxy carbonyl amino cyclohexanecarboxylic acid
285247
4-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
C1CC(CCC1C(=O)O)NC(=O)OCC2=CC=CC=C2
Quantity 1g
Melting Point 217°C
Formula Weight 277.31
Percent Purity 97%
Chemical Name or Material trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures