Learn More
Medchemexpress LLC trans-4-Hydroxycyclohexanecarboxylic acid | 3685-26-5 | 98.0% | C7H12O3 | 5 G
SDP
Supplier:  Medchemexpress LLC HY761995G
Encompass_Preferred
trans-4-Hydroxycyclohexanecarboxylic acid is a high-purity chemical compound, presenting as a white to off-white solid with a purity of 98.0%. Its chemical formula is C7H12O3 and its CAS number is 3685-26-5. This compound serves as a substrate in the production of cyclohexanecarboxylic acid. Furthermore, it is recognized as a by-product of intestinal bacterial metabolism, which is excreted via urine. With a molecular weight of 144.17, it is classified under ketones, aldehydes, and acids, and originates as an endogenous metabolite.
  • High-purity chemical compound
  • White to off-white solid appearance
  • Serves as a substrate in the production of cyclohexanecarboxylic acid
  • Recognized as a by-product of intestinal bacterial metabolism
  • Classified under ketones, aldehydes, and acids
  • Originates as an endogenous metabolite
  • Good solubility in DMSO, with a concentration of 100 mg/mL (693.63 mM)
Catalog No. 50-004-34186