Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Methoxycinnamaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
24680-50-0
C10H10O2
162.19
MFCD00017343
AXCXHFKZHDEKTP-NSCUHMNNSA-N
4-methoxycinnamaldehyde,3-4-methoxyphenyl acrylaldehyde,p-methoxycinnamaldehyde,e-3-4-methoxyphenyl acrylaldehyde,3-4-methoxyphenyl-2-propenal,trans-4-methoxycinnamaldehyde,p-methoxy cinnamaldehyde,p-methoxycinnamic aldehyde,trans-p-methoxycinnamaldehyde,cinnamaldehyde, p-methoxy
641294
CHEBI:27952
(E)-3-(4-methoxyphenyl)prop-2-enal
COC1=CC=C(C=C1)C=CC=O
24680-50-0
C10H10O2
162.19
MFCD00017343
AXCXHFKZHDEKTP-NSCUHMNNSA-N
4-methoxycinnamaldehyde,3-4-methoxyphenyl acrylaldehyde,p-methoxycinnamaldehyde,e-3-4-methoxyphenyl acrylaldehyde,3-4-methoxyphenyl-2-propenal,trans-4-methoxycinnamaldehyde,p-methoxy cinnamaldehyde,p-methoxycinnamic aldehyde,trans-p-methoxycinnamaldehyde,cinnamaldehyde, p-methoxy
641294
CHEBI:27952
(E)-3-(4-methoxyphenyl)prop-2-enal
COC1=CC=C(C=C1)C=CC=O
Boiling Point 145°C
Melting Point 55°C to 60°C
Quantity 100g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Orange-Yellow
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 162.19
Physical Form Crystalline Powder or Chunks
Percent Purity 98%
Chemical Name or Material trans-4-Methoxycinnamaldehyde
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning