Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Methoxycinnamaldehyde, 98+%, Thermo Scientific™

Click to view available options
Quantity:
10g
50g
24680-50-0
C10H10O2
162.19
MFCD00017343
AXCXHFKZHDEKTP-NSCUHMNNSA-N
4-methoxycinnamaldehyde, 3-4-methoxyphenyl acrylaldehyde, p-methoxycinnamaldehyde, e-3-4-methoxyphenyl acrylaldehyde, 3-4-methoxyphenyl-2-propenal, trans-4-methoxycinnamaldehyde, p-methoxy cinnamaldehyde, p-methoxycinnamic aldehyde, trans-p-methoxycinnamaldehyde, cinnamaldehyde, p-methoxy
641294
CHEBI:27952
(E)-3-(4-methoxyphenyl)prop-2-enal
COC1=CC=C(\C=C\C=O)C=C1
24680-50-0
C10H10O2
162.19
MFCD00017343
AXCXHFKZHDEKTP-NSCUHMNNSA-N
4-methoxycinnamaldehyde, 3-4-methoxyphenyl acrylaldehyde, p-methoxycinnamaldehyde, e-3-4-methoxyphenyl acrylaldehyde, 3-4-methoxyphenyl-2-propenal, trans-4-methoxycinnamaldehyde, p-methoxy cinnamaldehyde, p-methoxycinnamic aldehyde, trans-p-methoxycinnamaldehyde, cinnamaldehyde, p-methoxy
641294
CHEBI:27952
(E)-3-(4-methoxyphenyl)prop-2-enal
COC1=CC=C(\C=C\C=O)C=C1

trans-4-Methoxycinnamaldehyde is used as an intermediate in chemical research.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
trans-4-Methoxycinnamaldehyde is used as an intermediate in chemical research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Incompatible with strong oxidizing agents.
Melting Point 56°C to 60°C
Boiling Point 160°C (3mmHg)
Flash Point >110°C (230°F)
Odor Pleasant
Beilstein 1447540
Quantity 50g
Solubility Information Insoluble in water.
Sensitivity Air Sensitive
Formula Weight 162.19
Percent Purity ≥98%
Chemical Name or Material trans-4-Methoxycinnamaldehyde
Show More Show Less

GHS H Statement
H319
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-807-0

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only