Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ trans-4-n-Pentylcyclohexanecarboxylic acid, 98%

Click to view available options
Quantity:
1g
5g
38289-29-1
C12H22O2
198.306
MFCD00009964
RVLAXPQGTRTHEV-UHFFFAOYSA-N
trans-4-pentylcyclohexanecarboxylic acid,4-pentylcyclohexanecarboxylic acid,trans-4-n-pentylcyclohexanecarboxylic acid,cyclohexanecarboxylic acid, 4-pentyl,trans-4-amylcyclohexanecarboxylic acid,cyclohexanecarboxylic acid, 4-pentyl-, trans,4-n-pentylcyclohexane carboxylic acid,4-amylcyclohexanecarboxylic acid,rvlaxpqgtrthev-xypyzodxsa-n,1s,4r-4-pentylcyclohexane-1-carboxylic acid
98882
4-pentylcyclohexane-1-carboxylic acid
CCCCCC1CCC(CC1)C(=O)O
38289-29-1
C12H22O2
198.306
MFCD00009964
RVLAXPQGTRTHEV-UHFFFAOYSA-N
trans-4-pentylcyclohexanecarboxylic acid,4-pentylcyclohexanecarboxylic acid,trans-4-n-pentylcyclohexanecarboxylic acid,cyclohexanecarboxylic acid, 4-pentyl,trans-4-amylcyclohexanecarboxylic acid,cyclohexanecarboxylic acid, 4-pentyl-, trans,4-n-pentylcyclohexane carboxylic acid,4-amylcyclohexanecarboxylic acid,rvlaxpqgtrthev-xypyzodxsa-n,1s,4r-4-pentylcyclohexane-1-carboxylic acid
98882
4-pentylcyclohexane-1-carboxylic acid
CCCCCC1CCC(CC1)C(=O)O
Flash Point >110°C (230°F)
Melting Point 52°C to 55°C
Quantity 1g
Formula Weight 198.3
Percent Purity 98%
Chemical Name or Material trans-4-n-Pentylcyclohexanecarboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning