Alfa Aesar™ trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl, 97%

Manufacturer: Alfa Aesar™ H5574806

Quantity 5g

 View more versions of this product

Catalog No. AAH5574806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 131819-23-3
    Molecular Formula C21H29F3
    Formula Weight 338.46
    MDL Number MFCD09838998
    Synonym 1,2,3-Trifluoro-5-[(trans,trans)-4'-n-propylbicyclohexyl-4-yl]benzene
    Chemical Name or Material trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl

    Specifications

    Formula Weight 338.46
    Melting Point 63° to 64°C
    Quantity 5g
    Molecular Formula C21H29F3
    MDL Number MFCD09838998
    CAS 131819-23-3
    Synonym 1,2,3-Trifluoro-5-[(trans,trans)-4'-n-propylbicyclohexyl-4-yl]benzene
    Chemical Name or Material trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl
    Assay 97%

    Description

    Applications
    trans-4-n-Propyl-4 ft.-(3,4,5-trifluorophenyl)bicyclohexyl is used to make PDLC mixtures and Switchable Film . It is also mainly used as a liquid crystal monomer, liquid crystal material.

    Solubility
    It is soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit