Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Pentylcyclohexanecarboxylic Acid 99.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
38289-29-1
C12H22O2
198.306
MFCD00009964
RVLAXPQGTRTHEV-UHFFFAOYSA-N
trans-4-pentylcyclohexanecarboxylic acid, 4-pentylcyclohexanecarboxylic acid, trans-4-n-pentylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-pentyl, trans-4-amylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-pentyl-, trans, 4-n-pentylcyclohexane carboxylic acid, 4-amylcyclohexanecarboxylic acid, rvlaxpqgtrthev-xypyzodxsa-n, 1s,4r-4-pentylcyclohexane-1-carboxylic acid
98882
4-pentylcyclohexane-1-carboxylic acid
CCCCCC1CCC(CC1)C(=O)O
38289-29-1
C12H22O2
198.306
MFCD00009964
RVLAXPQGTRTHEV-UHFFFAOYSA-N
trans-4-pentylcyclohexanecarboxylic acid, 4-pentylcyclohexanecarboxylic acid, trans-4-n-pentylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-pentyl, trans-4-amylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-pentyl-, trans, 4-n-pentylcyclohexane carboxylic acid, 4-amylcyclohexanecarboxylic acid, rvlaxpqgtrthev-xypyzodxsa-n, 1s,4r-4-pentylcyclohexane-1-carboxylic acid
98882
4-pentylcyclohexane-1-carboxylic acid
CCCCCC1CCC(CC1)C(=O)O
Color White
Melting Point 55°C
Quantity 25g
Formula Weight 198.31
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (GC,T)
Chemical Name or Material trans-4-Pentylcyclohexanecarboxylic Acid

EINECSNumber : (3)-3723

TSCA : No