Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-5-Decen-1-ol, 96%, Thermo Scientific Chemicals

Catalog No. AAL1176806
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56578-18-8
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00056187
InChI Key WYPQHXVMNVEVEB-AATRIKPKSA-N
Synonym trans-5-decen-1-ol, e-5-decen-1-ol, trans-5-decenol, 5-decen-1-ol, 5e, 5-decen-1-ol, e, decan-5-en-1-ol, e-5-decenol, unii-m709v74ozv, 5-decen-1-ol, epa pesticide chemical code 078038
PubChem CID 5283292
IUPAC Name (E)-dec-5-en-1-ol
SMILES CCCC\C=C\CCCCO

Specifications

Boiling Point 113°C to 118°C (11 mmHg)
Flash Point 62°C (143°F)
Refractive Index 1.452
Quantity 5 g
Beilstein 1840630
Formula Weight 156.27
Percent Purity 96%
Chemical Name or Material trans-5-Decen-1-ol
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 260-267-6
RTECSNumber HE2096000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.