Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-trans-ACPD, Tocris Bioscience™
GREENER_CHOICE

Catalog No. p-4783082
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Group I and II mGlu agonist

Equimolecular mixture of (1S,3R)- and (1R,3S)-ACPD. Selective agonist for metabotropic glutamate receptors; active at both group I and group II mGlu receptors (EC50 values are 2, 15, 23 and ∽800 μM at mGluR2, mGluR1, mGluR5 and mGluR4 respectively).

(1S,3R)-ACPD and cis-ACPD also available.

Chemical Identifiers

CAS 67684-64-4
Molecular Formula C7H11NO4
Molecular Weight (g/mol) 173.168
InChI Key YFYNOWXBIBKGHB-FFWSUHOLSA-N
Synonym 1s,3s-1-aminocyclopentane-1,3-dicarboxylic acid, chembl29726, trans-acpd, cis-acpd, +/--trans-acpd, 1s, 3r-acpd, 1-amino-cis-cyclopentane-1,3-dicarboxylic acid, tocris-0186, tocris-0187, tocris-0284
PubChem CID 6604704
IUPAC Name (1S,3S)-1-aminocyclopentane-1,3-dicarboxylic acid
SMILES C1CC(CC1C(=O)O)(C(=O)O)N

Specifications

Quantity 10 mg
Formula Weight 173.17
Percent Purity >99%
Chemical Name or Material (±)-trans-ACPD
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.