Learn More
trans-beta-Styrylboronic acid, 97%, Thermo Scientific Chemicals
Description
trans-beta-Styrylboronic acid as reagent is used for palladium (Pd)-catalyzed Suzuki-Miyaura coupling reactions, diastereoselective synthesis via Pd-catalyzed Heck-Suzuki cascade reaction and rhodium (Rh)-catalyzed intramolecular amination of aryl azides. It is also used as reagent in preparation of optically active unsaturated amino acids by diastereoselective Petasis borono-Mannich reaction and amino alcohol dienes via Petasis 3-component reaction using Ru-catalyzed ring-closing metathesis and isomerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans-beta-Styrylboronic acid as reagent is used for palladium (Pd)-catalyzed Suzuki-Miyaura coupling reactions, diastereoselective synthesis via Pd-catalyzed Heck-Suzuki cascade reaction and rhodium (Rh)-catalyzed intramolecular amination of aryl azides. It is also used as reagent in preparation of optically active unsaturated amino acids by diastereoselective Petasis borono-Mannich reaction and amino alcohol dienes via Petasis 3-component reaction using Ru-catalyzed ring-closing metathesis and isomerization.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications
Melting Point | 98°C to 100°C |
Molecular Formula | C11H10N4O4 |
Quantity | 1 g |
Synonym | carbadox, unii-m2x04r2e2y, getroxel, mecadox, dsstox_cid_23913, dsstox_rid_80088, dsstox_gsid_43913, fortigro, karbadox czech, methyl n-e-1-hydroxy-4-oxidoquinoxalin-4-ium-2-ylidene methyl iminocarbamate |
InChI Key | BPMVRAQIQQEBLN-UHFFFAOYSA-N |
SMILES | COC(=O)N=NC=C1C=[N+](C2=CC=CC=C2N1O)[O-] |
IUPAC Name | methyl N-[(E)-(1-hydroxy-4-oxidoquinoxalin-4-ium-2-ylidene)methyl]iminocarbamate |
Molecular Weight (g/mol) | 262.23 |
PubChem CID | 5353472 |
Formula Weight | 147.97 |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.