Learn More
Thomson Instrument CompanySupplier Diversity Partner TRANS CAP INVERSION 1.6L 4/PK DFS Item
Manufacturer:  Thomson Instrument CompanySupplier Diversity Partner 9317084
Inversion Trans.Cap for Opt. Growth™ 1.6L Flask 2' Tubing to weld to 7/16” C-Flex 24, 4/Pack
Catalog No. 50-111-9998