Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ trans-Cinnamoyl chloride, 97%

Manufacturer:  Alfa Aesar™ A1201636

Quantity 500g

 View more versions of this product

Catalog No. AAA1201636


  Check Availability
Add to cart

Specifications

Description

Applications
The derivative of this compound, trans-Cinnamoylimidazole is involved in the synthesis of trans-Cinnamoyl-Papain.

Solubility
Reacts with water.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases. Moisture sensitive.
Description & Specifications

Chemical Identifiers

17082-09-6
166.61
UN3261
14,2301
trans-Cinnamoyl chloride
C9H7ClO
MFCD00000732
606265
trans-3-Phenylacryloyl chloride

Specifications

17082-09-6
166.61
UN3261
14,2301
35° to 37°C
500g
Pungent
97%
C9H7ClO
MFCD00000732
606265
trans-3-Phenylacryloyl chloride
256° to 258°C
>110°C (230°F)
trans-Cinnamoyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit