Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Crotonyl chloride, 90%, tech., ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
625-35-4
C4H5ClO
104.54
MFCD00000734
RJUIDDKTATZJFE-NSCUHMNNSA-N
crotonyl chloride,2-butenoyl chloride,crotonoyl chloride,e-but-2-enoyl chloride,e-2-butenoyl chloride,crotonic acid chloride,2e-but-2-enoyl chloride,2-butenoyl chloride, e,2-butenoyl chloride, 2e,trans-2-butenoyl chloride
643131
(E)-but-2-enoyl chloride
CC=CC(=O)Cl
625-35-4
C4H5ClO
104.54
MFCD00000734
RJUIDDKTATZJFE-NSCUHMNNSA-N
crotonyl chloride,2-butenoyl chloride,crotonoyl chloride,e-but-2-enoyl chloride,e-2-butenoyl chloride,crotonic acid chloride,2e-but-2-enoyl chloride,2-butenoyl chloride, e,2-butenoyl chloride, 2e,trans-2-butenoyl chloride
643131
(E)-but-2-enoyl chloride
CC=CC(=O)Cl
Boiling Point 120°C to 123°C
Specific Gravity 1.09
Flash Point 35°C
Density 1.0900g/mL
Refractive Index 1.4585 to 1.4615
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Color Colorless to Brown
Assay Percent Range 90%
Linear Formula CH3CH=CHCOCl
Beilstein 02, 411
Fieser 13,208
Solubility Information Solubility in water: decomposes
Formula Weight 104.54
Physical Form Liquid
Percent Purity 90%
Grade Technical
Chemical Name or Material trans-Crotonyl chloride, tech., 90%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 210-889-9

TSCA : TSCA