Alfa Aesar™ trans-Diamminedichloropalladium(II), Premion™, 99.95% (metals basis), Pd 49.9% min

Manufacturer: Alfa Aesar™ 1103704

Quantity 2g

 View more versions of this product

Catalog No. AA1103704

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14323-43-4
    Formula Weight 211.37
    MDL Number MFCD00011621
    UN Number UN3077
    Synonym Dichlorodiamminepalladium(II)
    Chemical Name or Material trans-Diamminedichloropalladium(II)
    Grade Premion™

    Specifications

    Physical Form Crystalline Powder
    Formula Weight 211.37
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P310-P301+P312-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Solubility Information Slightly soluble in water; Soluble in NH4OH; Insoluble in chloroform, acetone
    MDL Number MFCD00011621
    Density 2.5g/mL
    Synonym Dichlorodiamminepalladium(II)
    TSCA TSCA
    Color Yellow
    CAS 14323-43-4
    UN Number UN3077
    Health Hazard 1 Danger
    Quantity 2g
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    RTECS Number RT3560000
    Concentration Pd 49.9% min.
    Health Hazard 2 GHS H Statement
    H318-H302
    Causes serious eye damage.
    Harmful if swallowed.
    Chemical Name or Material trans-Diamminedichloropalladium(II)
    Assay 99.95%, (metals basis)
    Grade Premion™

    GHS H Statement
    H318-H302
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P280-P264-P305+P351+P338-P310-P301+P312-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit