Alfa Aesar™ trans-Diamminedichloroplatinum(II), Pt 64.5% min

Manufacturer: Alfa Aesar™ 1047206

Quantity 5g

 View more versions of this product

Catalog No. AA1047206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14913-33-8
    Linear Formula PtCl2(NH3)2
    Formula Weight 300.06
    MDL Number MFCD00011623
    Synonym Dichlorodiammineplatinum(II); Transplatin
    Chemical Name or Material trans-Diamminedichloroplatinum(II)

    Specifications

    Formula Weight 300.06
    Health Hazard 2 GHS H Statement
    H334-H341-H335-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of causing genetic defects.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280h-P262-P305+P351+P338-P308+P313
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or concerned: Get medical advice/attention.
    MDL Number MFCD00011623
    Health Hazard 1 Danger
    Odor Odorless
    Physical Form Crystalline
    CAS 14913-33-8
    Synonym Dichlorodiammineplatinum(II); Transplatin
    Linear Formula PtCl2(NH3)2
    Concentration Pt 64.5% min.
    Melting Point 340°C (dec.)
    Color Yellow
    Quantity 5g
    RTECS Number TP2455000
    Chemical Name or Material trans-Diamminedichloroplatinum(II)

    GHS H Statement
    H334-H341-H335-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of causing genetic defects.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280h-P262-P305+P351+P338-P308+P313
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or concerned: Get medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit