Alfa Aesar™ trans-Dibromobis(triphenylphosphine)palladium(II), Pd 13.4%

Hydrogenation

Manufacturer: Alfa Aesar™ 1370404

Quantity 2g

 View more versions of this product

Catalog No. AA1370404

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22180-53-6
    Formula Weight 790.8
    MDL Number MFCD00134175
    Chemical Name or Material trans-Dibromobis(triphenylphosphine)palladium(II)

    Specifications

    Formula Weight 790.8
    Physical Form Powder
    CAS 22180-53-6
    Quantity 2g
    Sensitivity Hygroscopic
    MDL Number MFCD00134175
    Concentration Pd 13.4%
    Solubility Information Soluble in chloroform, toluene and benzene
    Chemical Name or Material trans-Dibromobis(triphenylphosphine)palladium(II)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit