Alfa Aesar™ trans-Dichlorobis(triethylphosphine)palladium(II)

Coupling of C-C bonds

Manufacturer: Alfa Aesar™ 3982303

Quantity 1g

 View more versions of this product

Catalog No. AA3982303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 28425-04-9
    Formula Weight 413.63
    MDL Number MFCD00191831
    UN Number UN3464
    Chemical Name or Material trans-Dichlorobis(triethylphosphine)palladium(II)

    Specifications

    Formula Weight 413.63
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 28425-04-9
    Solubility Information Soluble in chloroform, toluene and benzene
    Quantity 1g
    Sensitivity Hygroscopic
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
    MDL Number MFCD00191831
    UN Number UN3464
    Health Hazard 1 Danger
    Chemical Name or Material trans-Dichlorobis(triethylphosphine)palladium(II)

    GHS H Statement
    H301-H311-H331-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit