Alfa Aesar™ trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min

Hydrogenation, hydrosilation, carbonylation, oxidation, and C-C bond formation

Manufacturer: Alfa Aesar™ 1049114

Quantity 25g

 View more versions of this product

Catalog No. AA1049114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13965-03-2
    Linear Formula PdCl2[P(C6H5)3]2
    Formula Weight 701.91
    MDL Number MFCD00009593
    Synonym Bis(triphenylphosphine)dichloropalladium(II)
    Chemical Name or Material trans-Dichlorobis(triphenylphosphine)palladium(II)

    Specifications

    Formula Weight 701.91
    Concentration Pd 14.0% min.
    Melting Point ≈310°C (dec.)
    Color Yellow
    MDL Number MFCD00009593
    CAS 13965-03-2
    Synonym Bis(triphenylphosphine)dichloropalladium(II)
    Physical Form Crystalline
    Linear Formula PdCl2[P(C6H5)3]2
    Quantity 25g
    Odor Odorless
    Sensitivity Hygroscopic
    RTECS Number RT3578000
    Chemical Name or Material trans-Dichlorobis(triphenylphosphine)palladium(II)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit