Alfa Aesar™ trans-Dichlorobis(triphenylphosphine)palladium(II), Premion™, 99.95% (metals basis), Pd 14.7% min

Manufacturer: Alfa Aesar™ 4124514

Quantity 25g

 View more versions of this product

Catalog No. AA4124514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13965-03-2
    Formula Weight 701.91
    MDL Number MFCD00009593
    Chemical Name or Material trans-Dichlorobis(triphenylphosphine)palladium(II)
    Grade Premion™

    Specifications

    Formula Weight 701.91
    Physical Form Powder
    Concentration Pd 14.7% min.
    Odor Odorless
    MDL Number MFCD00009593
    CAS 13965-03-2
    Melting Point ≈310°C (dec.)
    Color Yellow
    Quantity 25g
    Sensitivity Hygroscopic
    RTECS Number RT3578000
    Chemical Name or Material trans-Dichlorobis(triphenylphosphine)palladium(II)
    Assay 99.95%, (metals basis)
    Grade Premion™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit