Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Dichlorodiamineplatinum(II), 99.95%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
14913-33-8
Cl2H6N2Pt
300.046
MFCD00011623
LXZZYRPGZAFOLE-UHFFFAOYSA-L
trans-dichlorodiamineplatinum ii,azane; dichloroplatinum,h6cl2n2pt,ia call,ddp-h,platinol tn,cl.2h3n.pt,platinum ii chloride diamine,spectrum1502107,cisplatin jp17/usp/inn
5702198
CHEBI:27899
azane;dichloroplatinum
N.N.Cl[Pt]Cl
14913-33-8
Cl2H6N2Pt
300.046
MFCD00011623
LXZZYRPGZAFOLE-UHFFFAOYSA-L
trans-dichlorodiamineplatinum ii,azane; dichloroplatinum,h6cl2n2pt,ia call,ddp-h,platinol tn,cl.2h3n.pt,platinum ii chloride diamine,spectrum1502107,cisplatin jp17/usp/inn
5702198
CHEBI:27899
azane;dichloroplatinum
N.N.Cl[Pt]Cl
for ultra trace analysis
Assay Percent Range 99.95%
Linear Formula Pt(NH3)2Cl2
Formula Weight 300.05
Physical Form Fine Crystalline Powder
Grade Trace Metal
Packaging Glass bottle
Total Trace Metal Impurities 0.05% max.
Color Yellow
Quantity 250mg
Chemical Name or Material trans-Dichlorodiamineplatinum(II), 99.95%

GHS H Statement:
Suspected of causing cancer.
Suspected of causing genetic defects.
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 238-980-9

RTECSNumber : TP2455000