Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
trans-Ferulic acid Matrix Substance for MALDI-MS (HPLC), ≥99.0% (HPLC), MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 462785GF
Specifications
5 g | |
Matrix Substance for MALDI-MS | |
MFCD00004400 | |
194.18 | |
Matrix Substance for MALDI-MS | |
Powder |
126833-17-8 | |
HOC6H3(OCH3)CH=CHCO2 H | |
trans-4-Hydroxy-3-methoxycinnamic acid; Ferulic acid | |
≥99.0% (HPLC) | |
168°C to 172°C; 171°C to 175°C |
Safety and Handling
RTECSNumber : UD3365500
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction