Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-β-Methylstyrene, Stabilized 97%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass bottle
873-66-5
C9H10
118.18
MFCD00009280
QROGIFZRVHSFLM-QHHAFSJGSA-N
beta-methylstyrene,trans-beta-methylstyrene,1-phenylpropene,isoallylbenzene,propenylbenzene,trans-1-phenyl-1-propene,1-propenylbenzene,trans-propenylbenzene,1-phenyl-1-propene,benzene, 1-propenyl
252325
[(E)-prop-1-enyl]benzene
CC=CC1=CC=CC=C1
873-66-5
C9H10
118.18
MFCD00009280
QROGIFZRVHSFLM-QHHAFSJGSA-N
beta-methylstyrene,trans-beta-methylstyrene,1-phenylpropene,isoallylbenzene,propenylbenzene,trans-1-phenyl-1-propene,1-propenylbenzene,trans-propenylbenzene,1-phenyl-1-propene,benzene, 1-propenyl
252325
[(E)-prop-1-enyl]benzene
CC=CC1=CC=CC=C1
Flash Point 58°C
Boiling Point 175°C
Specific Gravity 0.911
Refractive Index 1.5480 to 1.55
Density 0.9110g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 10g
Stabilizer 0.01% BHT
Assay Percent Range 97%
Linear Formula C6H5CH=CHCH3
Beilstein 05, 481
Formula Weight 118.18
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material trans-β-Methylstyrene, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-848-0

RTECSNumber : DA8400500