Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ trans-N-Benzyloxycarbonyl-4-tert-butoxy-L-proline, 95%

Click to view available options
Quantity:
1g
5g
25g
85201-91-8
C17H23NO5
321.373
MFCD00153336
YCTNIMOEFJORHQ-KGLIPLIRSA-N
z-hyp tbu-oh,z-o-tert-butyl-l-4-hydroxyproline,2s,4r-4-tert-butoxy-1,2-pyrrolidinedicarboxylic acid 1-benzyl ester,2s,4r-4-tert-butoxy-pyrrolidine-1,2-dicarboxylic acid 1-benzyl ester,2s,4r-1-benzyloxy carbonyl-4-tert-butoxy pyrrolidine-2-carboxylic acid,2s,4r-4-2-methylpropan-2-yl oxy-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
6950951
(2S,4R)-4-[(2-methylpropan-2-yl)oxy]-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
CC(C)(C)OC1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
85201-91-8
C17H23NO5
321.373
MFCD00153336
YCTNIMOEFJORHQ-KGLIPLIRSA-N
z-hyp tbu-oh,z-o-tert-butyl-l-4-hydroxyproline,2s,4r-4-tert-butoxy-1,2-pyrrolidinedicarboxylic acid 1-benzyl ester,2s,4r-4-tert-butoxy-pyrrolidine-1,2-dicarboxylic acid 1-benzyl ester,2s,4r-1-benzyloxy carbonyl-4-tert-butoxy pyrrolidine-2-carboxylic acid,2s,4r-4-2-methylpropan-2-yl oxy-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
6950951
(2S,4R)-4-[(2-methylpropan-2-yl)oxy]-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
CC(C)(C)OC1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
Beilstein 5088264
Formula Weight 321.37
Physical Form Solid
Percent Purity 95%
Quantity 25g
Chemical Name or Material trans-N-Benzyloxycarbonyl-4-tert-butoxy-L-proline

storageNote1 : 2° to 8°C