Learn More
Medchemexpress LLC Trans-N-(tert-butoxycarbonyl)-4-hydroxy-L-proline | 13726-69-7 | MFCD00053370 | >98.0% | 231.25 g/mol | C10H17NO5 | 10 G

Supplier: Medchemexpress LLC HYI078110G
Boc-Hyp-OH (trans-N-(tert-butoxycarbonyl)-4-hydroxy-L-proline, CAS 13726-69-7) is a Boc-protected hydroxyproline derivative supplied as a solid for research use. It is used as a building block in peptide synthesis and can function as a linker in antibody-drug conjugate and PROTAC chemistry.
- Boc-protected hydroxyproline derivative.
- Used as a building block in peptide synthesis.
- Applicable as a linker in ADC and PROTAC chemistry.
- Molecular weight 231.25 g/mol.
- Chemical formula C10H17NO5.
- Purity >98.0% (HPLC).
- Provided as a 10 G solid for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.