Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Stilbene, 98%, Thermo Scientific Chemicals

Catalog Number AAA1023222
Click to view available options
Quantity:
100 g
500 g
103-30-0
C14H12
180.25
MFCD00064300
PJANXHGTPQOBST-VAWYXSNFSA-N
trans-stilbene, e-stilbene, stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, bibenzylidene, bibenzylidine, bibenzal, trans-1,2-diphenylethene, alpha,beta-diphenylethylene
638088
CHEBI:36007
(E)-stilbene
C1=CC=C(C=C1)C=CC2=CC=CC=C2
103-30-0
C14H12
180.25
MFCD00064300
PJANXHGTPQOBST-VAWYXSNFSA-N
trans-stilbene, e-stilbene, stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, bibenzylidene, bibenzylidine, bibenzal, trans-1,2-diphenylethene, alpha,beta-diphenylethylene
638088
CHEBI:36007
(E)-stilbene
C1=CC=C(C=C1)C=CC2=CC=CC=C2

trans-Stilbene is used in the manufacturing of dye lasers, optical brighteners, non-steroidal synthetic estrogens such as diethylstilbestrol, fosfestrol and dienestrol. It is used as a phosphor and a scintillator. It is also involved in transition metal catalyzed asymmetric epoxidation and dihydroxylation. Further, it is used as raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Stilbene is used in the manufacturing of dye lasers, optical brighteners, non-steroidal synthetic estrogens such as diethylstilbestrol, fosfestrol and dienestrol. It is used as a phosphor and a scintillator. It is also involved in transition metal catalyzed asymmetric epoxidation and dihydroxylation. Further, it is used as raw material in organic synthesis.

Solubility
Soluble in benzene and ether. Insoluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 122°C to 125°C
Density 1.14
Boiling Point 305°C to 307°C
Odor Odorless
Quantity 100 g
UN Number UN3077
Beilstein 1616740
Merck Index 14,8817
Solubility Information Soluble in benzene and ether. Insoluble in water.
Formula Weight 180.25
Percent Purity 98%
Chemical Name or Material trans-Stilbene
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 203-098-5
RTECSNumber WJ4926500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit