Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Stilbene 98.0+%, TCI America™

Catalog Number s009025g
Click to view available options
Quantity:
25 g
100 g
500 g
103-30-0
C14H12
180.25
MFCD00064300
PJANXHGTPQOBST-VAWYXSNFSA-N
trans-stilbene, e-stilbene, stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, bibenzylidene, bibenzylidine, bibenzal, trans-1,2-diphenylethene, alpha,beta-diphenylethylene
638088
CHEBI:36007
(E)-stilbene
C1=CC=C(C=C1)C=CC2=CC=CC=C2
103-30-0
C14H12
180.25
MFCD00064300
PJANXHGTPQOBST-VAWYXSNFSA-N
trans-stilbene, e-stilbene, stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, bibenzylidene, bibenzylidine, bibenzal, trans-1,2-diphenylethene, alpha,beta-diphenylethylene
638088
CHEBI:36007
(E)-stilbene
C1=CC=C(C=C1)C=CC2=CC=CC=C2
Melting Point 123°C
Color White
Boiling Point 307°C
Quantity 25 g
Formula Weight 180.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-Stilbene
RTECSNumber WJ4926500
TSCA Yes
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit