Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-1,4-Diphenyl-1,3-butadiene 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
538-81-8
C16H14
206.29
MFCD00004791
JFLKFZNIIQFQBS-FNCQTZNRSA-N
bistyryl,1,4-diphenyl-1,3-butadiene,1,4-diphenylbutadiene,trans,trans-1,4-diphenyl-1,3-butadiene,1,4-diphenylerythrene,distyryl,trans,trans-1,4-diphenylbuta-1,3-diene,1,4-diphenylbuta-1,3-diene,1,1'-1e,3e-buta-1,3-diene-1,4-diyldibenzene,1,3-butadiene, 1,4-diphenyl
641683
CHEBI:35100
[(1E,3E)-4-phenylbuta-1,3-dienyl]benzene
C1=CC=C(C=C1)C=CC=CC2=CC=CC=C2
538-81-8
C16H14
206.29
MFCD00004791
JFLKFZNIIQFQBS-FNCQTZNRSA-N
bistyryl,1,4-diphenyl-1,3-butadiene,1,4-diphenylbutadiene,trans,trans-1,4-diphenyl-1,3-butadiene,1,4-diphenylerythrene,distyryl,trans,trans-1,4-diphenylbuta-1,3-diene,1,4-diphenylbuta-1,3-diene,1,1'-1e,3e-buta-1,3-diene-1,4-diyldibenzene,1,3-butadiene, 1,4-diphenyl
641683
CHEBI:35100
[(1E,3E)-4-phenylbuta-1,3-dienyl]benzene
C1=CC=C(C=C1)C=CC=CC2=CC=CC=C2
Boiling Point 350°C
Infrared Spectrum Authentic
Quantity 25g
Melting Point 151°C to 154°C
Color White to Yellow
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula C6H5CH=CHCH=CHC6H5
Beilstein 05, 676
Fieser 01,337
Merck Index 15, 3350
Solubility Information Solubility in water: negligible
Formula Weight 206.29
Physical Form Crystals or Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material trans,trans-1,4-Diphenyl-1,3-butadiene, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 212-952-6

TSCA : TSCA