Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-2,4-Decadienal, 95%, Tech., ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
25152-84-5
C10H16O
152.24
MFCD00007007
JZQKTMZYLHNFPL-BLHCBFLLSA-N
2e,4e-deca-2,4-dienal,2,4-decadienal,e,e-2,4-decadienal,trans,trans-2,4-decadienal,2,4-decadienal, 2e,4e,2e,4e-decadienal,2e,4e-2,4-decadienal,2,4-decadienal, e,e,2-trans-4-trans-decadienal,ccris 4029
5283349
(2E,4E)-deca-2,4-dienal
CCCCCC=CC=CC=O
25152-84-5
C10H16O
152.24
MFCD00007007
JZQKTMZYLHNFPL-BLHCBFLLSA-N
2e,4e-deca-2,4-dienal,2,4-decadienal,e,e-2,4-decadienal,trans,trans-2,4-decadienal,2,4-decadienal, 2e,4e,2e,4e-decadienal,2e,4e-2,4-decadienal,2,4-decadienal, e,e,2-trans-4-trans-decadienal,ccris 4029
5283349
(2E,4E)-deca-2,4-dienal
CCCCCC=CC=CC=O
Specific Gravity 0.85
Flash Point 85°C
Boiling Point 114°C to 116°C (10.0mmHg)
Density 0.8500g/mL
Refractive Index 1.5140 to 1.518
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
Quantity 25g
Assay Percent Range 92% min. (main isomer) (GC)
Linear Formula CH3(CH2)4CH=CHCH=CHCHO
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 152.24
Physical Form Liquid
Percent Purity 95%
Grade Technical
Chemical Name or Material trans, trans-2, 4-Decadienal
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 246-668-9